Learn More
Fisherbrand™ Fabric ESD Chair - High Bench Height with Medium Back, Adjustable Arms, Chrome Foot Ring, and ESD Casters in ESD Fabric
Catalog No. 14000465 Shop All Fisherbrand Products
Catalog No. 14-000-465
Get 1 Extra Fisherbrand Chair When You Buy 4
View all promotions
Add to Cart
Add to Cart
Fabric ESD Chair with Medium Back is height for 42 in. work surfaces, features ergonomic adjustments and electro-static components such as fabric, self-braking casters, and arms. Chair with Medium Back is manufactured to ensure proper grounding for static sensitive applications.
  • Medium Back
  • Adjustable from 26.25 to 36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Autoclavable Not Autoclavable
With Lumbar Support Yes
Adjustable Yes
Arm Style 6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Base Material Polished Cast Aluminum
Certifications/Compliance ANSI/ESD-STM-12.1-2006
Adjustable (Height) 26.25 to 36 in.
Back Style Standard
Material Fabric
Seating Type High Bench Height
WARNING: Cancer - www.P65Warnings.ca.gov