Learn More
MSC FABRIC FR HEAD COVER
Catalog No. 5000782607
Encompass
This flame retardant fabric headcover is designed as an accessory for welding helmets, offering extended protection for the crown of the head, throat/neck, and ears during welding or grinding applications. It helps shield against UV light, sparks, and spatter.
  • Headcover type accessory
  • Black color
  • Constructed from durable, flame-retardant fabric
  • Compatible with G5-01, G5-02, and 9000/9100 Series welding helmets
  • Designed for easy attachment
  • Provides extended protection for head, neck, and ears
  • Protects against UV light, sparks, and spatter
Catalog No. 50-007-82607 Supplier MSC Supplier No. 18481630
This flame retardant fabric headcover is designed as an accessory for welding helmets, offering extended protection for the crown of the head, throat/neck, and ears during welding or grinding applications. It helps shield against UV light, sparks, and spatter.
  • Headcover type accessory
  • Black color
  • Constructed from durable, flame-retardant fabric
  • Compatible with G5-01, G5-02, and 9000/9100 Series welding helmets
  • Designed for easy attachment
  • Provides extended protection for head, neck, and ears
  • Protects against UV light, sparks, and spatter