Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1690993
Encompass_Preferred
Fabric mantle for 6.75" buchner funnel, to 400 degrees C internal operating temperature, 220W, 115V CSA
Catalog No. NC1690993 Supplier Glas Col LLC Supplier No. 100A O818
Fabric mantle for 6.75" buchner funnel, to 400 degrees C internal operating temperature, 220W, 115V CSA