Learn More
GENOVIS INC FabRICATOR Enzyme, Cysteine Protease which Digests Antibodies 2000 Units
Catalog No. NC0817077
Encompass
FabRICaTOR enzyme is supplied as lyophilized powder with no preservatives added and is available in four different pack sizes;
This item is not returnable. View return policy
Catalog No. NC0817077 Supplier GENOVIS INC Supplier No. A0FR1020
FabRICaTOR enzyme is supplied as lyophilized powder with no preservatives added and is available in four different pack sizes;