Learn More
GENOVIS INC FABRICATOR 8 STRIP WELL
Catalog No. NC0659019
Encompass
FabRICATOR, 8×100 Units Lyophilized FabRICATOR (IdeS) for digestion of 8 x 100 μg of antibody. Enzyme is reconstituted by addition of water. FabRICATOR (IdesS) is a unique protease that cleaves IgGs and Fc-fusions at one single amino acid position in the lower hinge region generating Fc and F(ab')2 fragments.
This item is not returnable. View return policy
Catalog No. NC0659019 Supplier GENOVIS INC Supplier No. A0FR1008
FabRICATOR, 8×100 Units Lyophilized FabRICATOR (IdeS) for digestion of 8 x 100 μg of antibody. Enzyme is reconstituted by addition of water. FabRICATOR (IdesS) is a unique protease that cleaves IgGs and Fc-fusions at one single amino acid position in the lower hinge region generating Fc and F(ab')2 fragments.