Learn More
GENOVIS INC FabRICATOR MagIC | 2mL
Catalog No. NC1867813
Encompass
FabRICATOR MagIC employs magnetic beads for fast automated processing of up to 96 antibodies in parallel. The popular cysteine protease FabRICATOR (IdeS) immobilized on magnetic agarose beads delivers rapid and consistent subunit generation for multiple antibodies in parallel, perfect for fast automated analysis of CQAs in a middle-level LC/MS workflow.
This item is not returnable. View return policy
Catalog No. NC1867813 Supplier GENOVIS INC Supplier No. A0FRM024
FabRICATOR MagIC employs magnetic beads for fast automated processing of up to 96 antibodies in parallel. The popular cysteine protease FabRICATOR (IdeS) immobilized on magnetic agarose beads delivers rapid and consistent subunit generation for multiple antibodies in parallel, perfect for fast automated analysis of CQAs in a middle-level LC/MS workflow.