Learn More
GENOVIS INC FabRICATOR® Z Immobilized Microspin 10 x 0.5 mg
Catalog No. NC4031256
Encompass
Immobilized enzyme for below hinge digestion of 10 x 0.5 mg mouse IgG2a and IgG3 in spin columns
Catalog No. NC4031256 Supplier GENOVIS INC Supplier No. A0FZ6050
Immobilized enzyme for below hinge digestion of 10 x 0.5 mg mouse IgG2a and IgG3 in spin columns