Learn More
GENOVIS INC FabRICATOR® Z Immobilized Microspin 2 x 0.5 mg
Catalog No. NC4031254
Encompass
Immobilized enzyme for below hinge digestion of 2 x 0.5 mg mouse IgG2a and IgG3 in spin columns
Catalog No. NC4031254 Supplier GENOVIS INC Supplier No. A0FZ6010
Immobilized enzyme for below hinge digestion of 2 x 0.5 mg mouse IgG2a and IgG3 in spin columns