Learn More
GENOVIS INC FabRICATOR® Z Immobilized Microspin 5 x 0.5 mg
Catalog No. NC4031252
Encompass
Immobilized enzyme for below hinge digestion of 5 x 0.5 mg mouse IgG2a and IgG3 in spin columns
Catalog No. NC4031252 Supplier GENOVIS INC Supplier No. A0FZ6025
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Immobilized enzyme for below hinge digestion of 5 x 0.5 mg mouse IgG2a and IgG3 in spin columns