Learn More
Avanti Polar Lipids 3α-hydroxy-7α,12α-di-((O-ß-D-maltosyl)-2-hydroxyethoxy)-cholane, 1429405-95-7, 200 mg
Catalog No. NC1584933
Encompass
Empirical Formula: (Hill Notation) C52H90O25, Molecular Weight: 1115.26, Purity >99%. Façade-EM is a two-dimensional amphiphile with hydrophobic and hydrophilic faces that has superior ability to stabilize integral membrane proteins. This new class of detergents represents a departure from traditional one-dimensional polar-head/nonpolar-tail molecules.
Catalog No. NC1584933 Supplier Avanti Polar Lipids Supplier No. 850522P200MG
Empirical Formula: (Hill Notation) C52H90O25, Molecular Weight: 1115.26, Purity >99%. Façade-EM is a two-dimensional amphiphile with hydrophobic and hydrophilic faces that has superior ability to stabilize integral membrane proteins. This new class of detergents represents a departure from traditional one-dimensional polar-head/nonpolar-tail molecules.