Learn More
S-Curve™ Face Mask Dispenser
Catalog No. 19999819 Shop All S-Curve Technologies Products
Change view
Click to view available options
Dimensions (L x W x H):
10 x 10 x 6 in.
Catalog No. Dimensions (L x W x H)
19-999-819 10 x 10 x 6 in.
1 options
Catalog No. 19-999-819 Supplier S-Curve™ Supplier No. BD864PETG
Add to Cart
Add to Cart
Face mask dispenser with front access. It can be wall mounted and the PETG material is chemical and impact resistant.
A smaller dispenser designed to hold face masks, beard covers, hair nets, or bouffants.
  • Front access hole
  • 3/16 in. thick PETG material (PETG is safe with IPA)
  • 10 x 10 x 6 in.
  • Flame treated edges for a smooth finish
  • Keyholes on back for wall mounting
  • 1 per case
Specifications
Product Type Face Mask Dispenser
Dimensions (L x W x H) 10 x 10 x 6 in.