Learn More
DuPont™ Sierra™ Face Mask with Earloops
Catalog No. 190882117 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Ear loops
Catalog No.
19-088-2117 Ear loops
1 options
This item is not returnable. View return policy
Catalog No. 19-088-2117 Supplier DuPont™ Supplier No. ML7330BU000500BH
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Polypropylene face mask combines high breathability with exceptional filtration efficiency.
  • Mask is 7 in. size with earloops
  • Pleated polypropylene
  • Metal nose piece
  • One size fits most
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Mask
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Universal
Sterility Non-sterile
Style Pleated
Filter Efficiency >95% (BFE)
Includes Metal Nose Piece
Model ML7330
Series Sierra™