Learn More
Grainger Face Mill FM 200090RH FM 200 Dia
Catalog No. 502764258
Encompass_Preferred
The KYOCERA Indexable Face Mill is a 2 inch diameter face milling cutter with 4 indexable carbide inserts. It has a 1 inch shank diameter and comes with 4 APKT1604PDTR-GM inserts. The cutter body is made of steel and can mill a variety of ferrous and non-ferrous materials. With its 4 cutting edges, this face mill is well suited for light to medium duty face milling operations where a balance of economy and performance is required. The APKT inserts provide good chip control and are reversible, doubling the insert life. Overall, a solid face mill for general purpose face milling.
Catalog No. 50-276-4258 Supplier Grainger Supplier No. 170JZ6
The KYOCERA Indexable Face Mill is a 2 inch diameter face milling cutter with 4 indexable carbide inserts. It has a 1 inch shank diameter and comes with 4 APKT1604PDTR-GM inserts. The cutter body is made of steel and can mill a variety of ferrous and non-ferrous materials. With its 4 cutting edges, this face mill is well suited for light to medium duty face milling operations where a balance of economy and performance is required. The APKT inserts provide good chip control and are reversible, doubling the insert life. Overall, a solid face mill for general purpose face milling.