Learn More
Grainger Face Mill FM 300030RH FM 300 Dia
Catalog No. 502764165
Encompass_Preferred
The Kyocera Face Mill is a high-performance milling cutter for face milling, slotting and plunging. It has a diameter of 2 inches and an overall length of 3 inches. The face mill features 4 carbide inserts with a TiAIN coating for wear resistance when machining steel, stainless steel and cast iron. It has Weldon flat shank arbors available in multiple sizes for mounting in a milling machine. The maximum speed is 6,000 RPM.
Catalog No. 50-276-4165 Supplier Grainger Supplier No. 170JZ8
The Kyocera Face Mill is a high-performance milling cutter for face milling, slotting and plunging. It has a diameter of 2 inches and an overall length of 3 inches. The face mill features 4 carbide inserts with a TiAIN coating for wear resistance when machining steel, stainless steel and cast iron. It has Weldon flat shank arbors available in multiple sizes for mounting in a milling machine. The maximum speed is 6,000 RPM.