Learn More
Grainger Face Mill FM 300090AL FM-AL 300 Dia
Catalog No. 502764651
Encompass_Preferred
The Kyocera Indexable Face Mill is a 3" diameter face milling cutter with indexable carbide inserts. It has 4 cutting edges and uses TPG432 style inserts. The cutter body is made of steel with a TiAIN coating and has a 1" arbor hole. It is capable of light to medium duty facing, surfacing and slotting applications in steel, stainless steel, cast iron and non-ferrous metals. The maximum depth of cut is 0.315" and maximum feed rate is 0.004" per tooth. It has coolant holes to allow high pressure coolant to aid chip removal. The cutter comes with 4 inserts and a removal tool for changing inserts.
Catalog No. 50-276-4651 Supplier Grainger Supplier No. 170JZ9
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Kyocera Indexable Face Mill is a 3" diameter face milling cutter with indexable carbide inserts. It has 4 cutting edges and uses TPG432 style inserts. The cutter body is made of steel with a TiAIN coating and has a 1" arbor hole. It is capable of light to medium duty facing, surfacing and slotting applications in steel, stainless steel, cast iron and non-ferrous metals. The maximum depth of cut is 0.315" and maximum feed rate is 0.004" per tooth. It has coolant holes to allow high pressure coolant to aid chip removal. The cutter comes with 4 inserts and a removal tool for changing inserts.