Learn More
Grainger Face Mill FM 300090RH FM 300 Dia
Catalog No. 502764297
Encompass_Preferred
The Kyocera Indexable Face Mill is a 3" diameter face milling cutter with a maximum depth of cut of 0.315". It has 4 indexable carbide inserts with a TiAlN coating for wear resistance. The cutter body is made of steel and has a BT30 toolholder shank. It is capable of face milling, slotting and side milling operations. The maximum spindle speed is 4000 RPM.
Catalog No. 50-276-4297 Supplier Grainger Supplier No. 170KA0
The Kyocera Indexable Face Mill is a 3" diameter face milling cutter with a maximum depth of cut of 0.315". It has 4 indexable carbide inserts with a TiAlN coating for wear resistance. The cutter body is made of steel and has a BT30 toolholder shank. It is capable of face milling, slotting and side milling operations. The maximum spindle speed is 4000 RPM.