Learn More
Grainger Face Mill FM 400090AL125 FM-AL 400
Catalog No. 502764429
Encompass_Preferred
The Kyocera Indexable Face Mill is a 4" diameter face milling cutter with a maximum depth of cut of 0.5". It has 4 indexable carbide inserts with a TiAlN coating for wear resistance. The cutter body is made of steel and has a BT30 toolholder shank. It is capable of face milling, slotting and side milling operations. The maximum spindle speed is 4000 RPM.
Catalog No. 50-276-4429 Supplier Grainger Supplier No. 170KA2
The Kyocera Indexable Face Mill is a 4" diameter face milling cutter with a maximum depth of cut of 0.5". It has 4 indexable carbide inserts with a TiAlN coating for wear resistance. The cutter body is made of steel and has a BT30 toolholder shank. It is capable of face milling, slotting and side milling operations. The maximum spindle speed is 4000 RPM.