Learn More
Grainger Face Mill FMAL 4000905 FM-AL 400 Dia
Catalog No. 502764430
Encompass_Preferred
The KYOCERA Indexable Face Mill is a 4" diameter face milling cutter with a maximum depth of cut of 0.630". It has 4 indexable carbide inserts with a TPG432 style insert and uses a Weldon style shank. The overall length is 4.72" and it has a 1.25" through coolant hole. The body material is steel with a black oxide finish. This face mill is designed for general purpose face milling in steel and cast iron. It has high feed rates and can provide excellent surface finishes. The replaceable carbide inserts allow for economical operation.
Catalog No. 50-276-4430 Supplier Grainger Supplier No. 170KA5
The KYOCERA Indexable Face Mill is a 4" diameter face milling cutter with a maximum depth of cut of 0.630". It has 4 indexable carbide inserts with a TPG432 style insert and uses a Weldon style shank. The overall length is 4.72" and it has a 1.25" through coolant hole. The body material is steel with a black oxide finish. This face mill is designed for general purpose face milling in steel and cast iron. It has high feed rates and can provide excellent surface finishes. The replaceable carbide inserts allow for economical operation.