Learn More
Grainger Face Mill MFPN 453000R6T MFPN 300
Catalog No. 502764305
Encompass_Preferred
The Kyocera Face Mill is a high-performance cutter for high-speed machining of aluminum and non-ferrous metals. It has a 1" diameter, 4 flute design with replaceable carbide inserts for a superior finish. Made of heat-treated alloy steel, it offers rigidity and durability. With a maximum speed of 12,000 RPM and feed rates up to 0.004" per tooth, it includes 5 carbide inserts with TiAlN coating for wear resistance. Ideal for face milling, slotting, and side milling in aerospace and automotive industries, this versatile face mill excels in demanding operations.
Catalog No. 50-276-4305 Supplier Grainger Supplier No. 170KC0
The Kyocera Face Mill is a high-performance cutter for high-speed machining of aluminum and non-ferrous metals. It has a 1" diameter, 4 flute design with replaceable carbide inserts for a superior finish. Made of heat-treated alloy steel, it offers rigidity and durability. With a maximum speed of 12,000 RPM and feed rates up to 0.004" per tooth, it includes 5 carbide inserts with TiAlN coating for wear resistance. Ideal for face milling, slotting, and side milling in aerospace and automotive industries, this versatile face mill excels in demanding operations.