Learn More
Grainger Face Mill MFPN 455000R13T MFPN 500
Catalog No. 502764449
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches and an overall length of 3 inches. It has 4 indexable carbide inserts with a TiAIN coating for wear resistance when milling steel, stainless steel, cast iron, and other hard materials. The cutter body is made of steel and features a Weldon flat shank with a 7/8 inch shank diameter to fit milling machines. The maximum depth of cut is 1 inch and it has 4 flutes for efficient chip removal. With its rigid design and sharp cutting edges, this face mill is well-suited for heavy roughing and finishing operations.
Catalog No. 50-276-4449 Supplier Grainger Supplier No. 170KC4
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches and an overall length of 3 inches. It has 4 indexable carbide inserts with a TiAIN coating for wear resistance when milling steel, stainless steel, cast iron, and other hard materials. The cutter body is made of steel and features a Weldon flat shank with a 7/8 inch shank diameter to fit milling machines. The maximum depth of cut is 1 inch and it has 4 flutes for efficient chip removal. With its rigid design and sharp cutting edges, this face mill is well-suited for heavy roughing and finishing operations.