Learn More
Grainger Face Mill MFPN 456000R12T MFPN 600
Catalog No. 502763823
Encompass_Preferred
The Kyocera Face Mill is a high-performance cutter for high-speed machining of steel, stainless steel, cast iron, and non-ferrous metals. It has a 2-flute design with replaceable carbide inserts for smooth cutting and long tool life. Made of high-quality tool steel, it features a 1" arbor hole and diameters from 1" to 4". It achieves surface finishes as fine as 32 microinches Ra. The high-precision insert seat and rigid clamping system ensure accurate depth of cut and prevent insert movement. Suitable for facing, slotting, side milling, and contouring operations.
Catalog No. 50-276-3823 Supplier Grainger Supplier No. 170KC1
The Kyocera Face Mill is a high-performance cutter for high-speed machining of steel, stainless steel, cast iron, and non-ferrous metals. It has a 2-flute design with replaceable carbide inserts for smooth cutting and long tool life. Made of high-quality tool steel, it features a 1" arbor hole and diameters from 1" to 4". It achieves surface finishes as fine as 32 microinches Ra. The high-precision insert seat and rigid clamping system ensure accurate depth of cut and prevent insert movement. Suitable for facing, slotting, side milling, and contouring operations.