Learn More
Grainger Face Mill MFPN 456000R8T MFPN 600
Catalog No. 502764447
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches and an overall length of 3 inches. It has 4 indexable carbide inserts with a TiAIN PVD coating for wear resistance. The cutter body is made of steel and has a right-hand spiral flute design. The maximum depth of cut is 0.630 inches and it has a shank diameter of 0.750 inches. The cutter is designed for high feed milling of steel, stainless steel, cast iron, and other ferrous metals. It provides accurate 90 degree shoulder milling and facing operations.
Catalog No. 50-276-4447 Supplier Grainger Supplier No. 170KA8
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches and an overall length of 3 inches. It has 4 indexable carbide inserts with a TiAIN PVD coating for wear resistance. The cutter body is made of steel and has a right-hand spiral flute design. The maximum depth of cut is 0.630 inches and it has a shank diameter of 0.750 inches. The cutter is designed for high feed milling of steel, stainless steel, cast iron, and other ferrous metals. It provides accurate 90 degree shoulder milling and facing operations.