Learn More
Grainger Face Mill MFPN 458000R10T MFPN 800
Catalog No. 502764199
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter for machining steel, cast iron, and non-ferrous metals. It features a 2.5" diameter, 4 flute design with a 1" arbor hole and a 1.5" depth of cut. The cutter is made of micrograin carbide with a TiAlN coating for wear resistance and heat resistance. It has a 45 degree lead angle and can achieve surface finishes up to 32 microinches. The cutter is designed for high feed milling applications with feed rates up to 0.012" per tooth. It is well-suited for facing, slotting, and surfacing in high production environments. The Kyocera Face Mill combines durability, performance, and versatility for demanding metal removal applications.
Catalog No. 50-276-4199 Supplier Grainger Supplier No. 170KA9
The Kyocera Face Mill is a high-performance face milling cutter for machining steel, cast iron, and non-ferrous metals. It features a 2.5" diameter, 4 flute design with a 1" arbor hole and a 1.5" depth of cut. The cutter is made of micrograin carbide with a TiAlN coating for wear resistance and heat resistance. It has a 45 degree lead angle and can achieve surface finishes up to 32 microinches. The cutter is designed for high feed milling applications with feed rates up to 0.012" per tooth. It is well-suited for facing, slotting, and surfacing in high production environments. The Kyocera Face Mill combines durability, performance, and versatility for demanding metal removal applications.