Learn More
Grainger Face Mill MFWN 902500R4T M-SIX 2.5000
Catalog No. 502764277
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches. It features 4 indexable carbide inserts with a TiAIN coating for wear resistance when milling steel, stainless steel, cast iron, and other materials. The cutter has 4 flutes and can achieve optimal chip control and evacuation. With its 45 degree lead angle inserts, this face mill is well-suited for light to heavy milling applications. The cutter body is made of steel and includes a Weldon flat and through coolant holes. The maximum speed is 11,000 RPM.
Catalog No. 50-276-4277 Supplier Grainger Supplier No. 170KC8
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches. It features 4 indexable carbide inserts with a TiAIN coating for wear resistance when milling steel, stainless steel, cast iron, and other materials. The cutter has 4 flutes and can achieve optimal chip control and evacuation. With its 45 degree lead angle inserts, this face mill is well-suited for light to heavy milling applications. The cutter body is made of steel and includes a Weldon flat and through coolant holes. The maximum speed is 11,000 RPM.