Learn More
Grainger Face Mill MFWN 902500R5T M-SIX 2.5000
Catalog No. 502764299
Encompass_Preferred
The Kyocera Face Mill is a high-performance milling cutter for face milling various materials. It has a 45-degree lead angle and variable helix flutes for efficient chip evacuation. Made of micro-grain carbide, it offers wear resistance and durability. With 6 indexable carbide inserts coated with TiAlN, it ensures hardness and heat resistance. Available in diameters from 1" to 6", it runs smoothly at high RPMs. Its rigid design and sharp edges provide excellent surface finishes and accuracy, ideal for heavy roughing and finishing in automotive, aerospace, die/mold, and general machining.
Catalog No. 50-276-4299 Supplier Grainger Supplier No. 170KD1
The Kyocera Face Mill is a high-performance milling cutter for face milling various materials. It has a 45-degree lead angle and variable helix flutes for efficient chip evacuation. Made of micro-grain carbide, it offers wear resistance and durability. With 6 indexable carbide inserts coated with TiAlN, it ensures hardness and heat resistance. Available in diameters from 1" to 6", it runs smoothly at high RPMs. Its rigid design and sharp edges provide excellent surface finishes and accuracy, ideal for heavy roughing and finishing in automotive, aerospace, die/mold, and general machining.