Learn More
Grainger Face Mill MFWN 903000R4T M-SIX 300
Catalog No. 502764191
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter designed for high-speed machining of aluminum and non-ferrous metals. It features a 2-flute design with replaceable carbide inserts that provide a smooth finish and long tool life. The cutter body is made of heat-treated alloy steel for rigidity and durability. It has a 1" arbor hole and is available in diameters ranging from 1" to 4". The cutter can achieve surface speeds up to 13,000 SFM and feed rates up to 0.004 IPR. With its rigid construction and sharp cutting edges, this face mill is ideal for facing, slotting, side milling, and contouring operations where an excellent finish is required.
Catalog No. 50-276-4191 Supplier Grainger Supplier No. 170KC5
The Kyocera Face Mill is a high-performance face milling cutter designed for high-speed machining of aluminum and non-ferrous metals. It features a 2-flute design with replaceable carbide inserts that provide a smooth finish and long tool life. The cutter body is made of heat-treated alloy steel for rigidity and durability. It has a 1" arbor hole and is available in diameters ranging from 1" to 4". The cutter can achieve surface speeds up to 13,000 SFM and feed rates up to 0.004 IPR. With its rigid construction and sharp cutting edges, this face mill is ideal for facing, slotting, side milling, and contouring operations where an excellent finish is required.