Learn More
Grainger Face Mill MFWN 903000R5T M-SIX 300
Catalog No. 502764298
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter for machining steel, cast iron, and non-ferrous metals. It features a 2.5" diameter with 4 carbide inserts for a maximum depth of cut of 0.5". The cutter has a 45 degree lead angle and is designed for high feed rates and smooth milling action. The carbide inserts utilize a PVD coating for increased tool life and heat resistance. The cutter body is made of heat-treated steel for rigidity. Recommended cutting speeds range from 400-1000 SFM based on workpiece material.
Catalog No. 50-276-4298 Supplier Grainger Supplier No. 170KC9
The Kyocera Face Mill is a high-performance face milling cutter for machining steel, cast iron, and non-ferrous metals. It features a 2.5" diameter with 4 carbide inserts for a maximum depth of cut of 0.5". The cutter has a 45 degree lead angle and is designed for high feed rates and smooth milling action. The carbide inserts utilize a PVD coating for increased tool life and heat resistance. The cutter body is made of heat-treated steel for rigidity. Recommended cutting speeds range from 400-1000 SFM based on workpiece material.