Learn More
Grainger Face Mill MFWN 904000R5T M-SIX 400
Catalog No. 502764450
Encompass_Preferred
The Kyocera Face Mill is a face milling cutter with a 2.5" diameter and a 1" arbor hole. It has 4 indexable carbide inserts with a TiAlN coating for wear resistance. The inserts have 8 cutting edges for extended tool life. The cutter body is made of steel and can mill a variety of ferrous and non-ferrous metals. With its 4 flutes, it can provide efficient chip evacuation when face milling. This versatile face mill is well-suited for high speed machining applications.
Catalog No. 50-276-4450 Supplier Grainger Supplier No. 170KC6
The Kyocera Face Mill is a face milling cutter with a 2.5" diameter and a 1" arbor hole. It has 4 indexable carbide inserts with a TiAlN coating for wear resistance. The inserts have 8 cutting edges for extended tool life. The cutter body is made of steel and can mill a variety of ferrous and non-ferrous metals. With its 4 flutes, it can provide efficient chip evacuation when face milling. This versatile face mill is well-suited for high speed machining applications.