Learn More
Grainger Face Mill MFWN 904000R7T M-SIX 400
Catalog No. 502764385
Encompass_Preferred
The 170KD0 is a Dayton 1/3 HP General Purpose Motor made by Dayton Electric Manufacturing. It is a single phase AC motor with a totally enclosed fan cooled (TEFC) enclosure and operates on 115V at 60Hz. The motor has a 1/3 HP rating with a 1725 RPM speed and 48Y frame. It features a cast iron body with pre-lubricated sleeve bearings and automatic reset thermal overload protection. The motor dimensions are 7-1/4" diameter x 10-3/4" long and it weighs 26 lbs. This electric motor is suitable for various industrial, commercial and residential applications.
Catalog No. 50-276-4385 Supplier Grainger Supplier No. 170KD0
The 170KD0 is a Dayton 1/3 HP General Purpose Motor made by Dayton Electric Manufacturing. It is a single phase AC motor with a totally enclosed fan cooled (TEFC) enclosure and operates on 115V at 60Hz. The motor has a 1/3 HP rating with a 1725 RPM speed and 48Y frame. It features a cast iron body with pre-lubricated sleeve bearings and automatic reset thermal overload protection. The motor dimensions are 7-1/4" diameter x 10-3/4" long and it weighs 26 lbs. This electric motor is suitable for various industrial, commercial and residential applications.