Learn More
Grainger Face Mill MFWN 906000R8T M-SIX 600
Catalog No. 502764431
Encompass_Preferred
The Kyocera Face Mill is a high-performance milling cutter designed for high-speed machining of aluminum, steel, cast iron, and other materials. It features a 2-flute design with replaceable carbide inserts that provide a smooth finish and long tool life. The cutter has a 1" shank diameter and is available in diameters from 1" to 4" with a maximum depth of cut up to 1". It can achieve surface speeds up to 6600 SFM and feed rates up to 0.004 IPR. The cutter body is made of heat-treated, forged alloy steel for strength and rigidity. Overall, this face mill is well-suited for heavy roughing and finishing operations where smoothness, speed, and durability are required.
Catalog No. 50-276-4431 Supplier Grainger Supplier No. 170KC7
The Kyocera Face Mill is a high-performance milling cutter designed for high-speed machining of aluminum, steel, cast iron, and other materials. It features a 2-flute design with replaceable carbide inserts that provide a smooth finish and long tool life. The cutter has a 1" shank diameter and is available in diameters from 1" to 4" with a maximum depth of cut up to 1". It can achieve surface speeds up to 6600 SFM and feed rates up to 0.004 IPR. The cutter body is made of heat-treated, forged alloy steel for strength and rigidity. Overall, this face mill is well-suited for heavy roughing and finishing operations where smoothness, speed, and durability are required.