Learn More
Waters Corp Waters Corporation Face Seals Replacement Kit, 4 pk, Pump Head, HPLC System
Catalog No. 502039977 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides the necessary components for maintaining the performance of your system. It contains four face seals, enabling the replacement of seals on both pump heads in the Alliance 2695 module, ensuring continued reliable operation.
  • Component compatibility: 2695 Separations Module and 2695 D Dissolution Separations Module.
  • Units per package: 4.
  • Designed for pump heads.
  • Suitable for HPLC systems.
  • Recommended annual replacement.
Catalog No. 50-203-9977 Supplier Waters Corp Supplier No. WAT270939
This kit provides the necessary components for maintaining the performance of your system. It contains four face seals, enabling the replacement of seals on both pump heads in the Alliance 2695 module, ensuring continued reliable operation.
  • Component compatibility: 2695 Separations Module and 2695 D Dissolution Separations Module.
  • Units per package: 4.
  • Designed for pump heads.
  • Suitable for HPLC systems.
  • Recommended annual replacement.