Learn More
MSC Sellstrom Face Shield with Chin Guard, Polycarbonate, 1 ea
Catalog No. NC1833904
Encompass
This face shield with chin guard provides protection for industrial and laboratory environments. Constructed from polycarbonate, it features a clear, scratch-resistant window and a blue nylon headgear with ratchet adjustment for a secure fit. It meets CE, CSA Z94.3, and ANSI Z87.1+ safety standards.
  • Polycarbonate window material
  • Scratch-resistant window coating
  • Window thickness 0.0400 in
  • Window height 6-1/2 in, width 19-1/2 in
  • Blue nylon headgear
  • Ratchet adjustment
  • Extra-large, extra-wide design
  • Includes two temple adjustments
  • Meets CE, CSA Z94.3, ANSI Z87.1+ standards
  • Made in the United States
This item is not returnable. View return policy
Catalog No. NC1833904 Supplier MSC Supplier No. 90402587
This face shield with chin guard provides protection for industrial and laboratory environments. Constructed from polycarbonate, it features a clear, scratch-resistant window and a blue nylon headgear with ratchet adjustment for a secure fit. It meets CE, CSA Z94.3, and ANSI Z87.1+ safety standards.
  • Polycarbonate window material
  • Scratch-resistant window coating
  • Window thickness 0.0400 in
  • Window height 6-1/2 in, width 19-1/2 in
  • Blue nylon headgear
  • Ratchet adjustment
  • Extra-large, extra-wide design
  • Includes two temple adjustments
  • Meets CE, CSA Z94.3, ANSI Z87.1+ standards
  • Made in the United States