Learn More
Hach Company Face Shield, Polycarbonate
Supplier:  Hach Company 2928101
Encompass
Fully adjustable head straps with padded brow provides comfort, while 15" wide x 8 1/2" tall polycarbonate shield provides outstanding splash protection. Shield also provides 99.9% UV protection up to 380nm. Forehead cover and ledge design prevents fluids from splashing inside the window. Removable, steam autoclavable face shield. Meets ANSI Z87+ and CSA Z94.3 standards. Some Assembly Required.
Catalog No. 50-199-6805