Learn More
Grainger Fibre-Metal By Pip Face Shield System, Propionate, Clear, 1 ea
Catalog No. 502761244
Encompass_Preferred
This face shield system provides lightweight, comfortable, and reliable protection for general-purpose applications. The assembly includes a clear propionate visor, a gray 3C ratchet headgear, and a chin guard, offering comprehensive wraparound facial coverage.
  • Features an extended, wraparound design for enhanced protection
  • Equipped with an integrated crown and chin guard for complete coverage
  • Utilizes a 3C ratchet suspension system for secure and customizable fit
  • Dielectric construction suitable for electrical work environments
  • Comes with a clear, uncoated, reusable propionate window
  • Meets ANSI Z87.1-2003 safety standards
Catalog No. 50-276-1244 Supplier Grainger Supplier No. 15W978
This face shield system provides lightweight, comfortable, and reliable protection for general-purpose applications. The assembly includes a clear propionate visor, a gray 3C ratchet headgear, and a chin guard, offering comprehensive wraparound facial coverage.
  • Features an extended, wraparound design for enhanced protection
  • Equipped with an integrated crown and chin guard for complete coverage
  • Utilizes a 3C ratchet suspension system for secure and customizable fit
  • Dielectric construction suitable for electrical work environments
  • Comes with a clear, uncoated, reusable propionate window
  • Meets ANSI Z87.1-2003 safety standards