Learn More
Grainger Fibre-Metal By Pip Face Shield Assembly, Propionate, Clear, Uncoated, 16 1/2 in. Visor, 1 ea
Catalog No. 502760814
Encompass_Preferred
This general purpose face shield assembly features a single-crown design with an integrated chin guard to provide comprehensive protection against splashes and airborne debris. The crown protects the forehead and top of the head, while the chin guard enhances stability and shields the underside of the chin. Constructed from durable propionate, the clear visor offers high visibility and includes a ratchet suspension for secure, adjustable wear.
  • Clear, uncoated propionate visor with 16-1/2 in width
  • Integrated chin guard for added stability and underside protection
  • Ratchet suspension system for customizable fit
  • Dielectric construction
  • Meets ANSI Z87.1-2003 standards
  • Reusable design
Catalog No. 50-276-0814 Supplier Grainger Supplier No. 15W976
This general purpose face shield assembly features a single-crown design with an integrated chin guard to provide comprehensive protection against splashes and airborne debris. The crown protects the forehead and top of the head, while the chin guard enhances stability and shields the underside of the chin. Constructed from durable propionate, the clear visor offers high visibility and includes a ratchet suspension for secure, adjustable wear.
  • Clear, uncoated propionate visor with 16-1/2 in width
  • Integrated chin guard for added stability and underside protection
  • Ratchet suspension system for customizable fit
  • Dielectric construction
  • Meets ANSI Z87.1-2003 standards
  • Reusable design