Learn More
Grainger North By Honeywell Faceshield Bracket, Aluminum, High Heat Resistant, 1 Ea.
Catalog No. 502761503
Encompass_Preferred
These aluminum brackets are designed for high heat resistance, providing reliable face shield support for various head protection applications. Constructed from durable aluminum, they ensure robust performance in demanding environments.
  • High heat resistant construction
  • Compatible with most standard cap style hard hats
  • Fits North and various competitive hard hat models
  • Designed for use with face shields
Catalog No. 50-276-1503 Supplier Grainger Supplier No. 15W991
These aluminum brackets are designed for high heat resistance, providing reliable face shield support for various head protection applications. Constructed from durable aluminum, they ensure robust performance in demanding environments.
  • High heat resistant construction
  • Compatible with most standard cap style hard hats
  • Fits North and various competitive hard hat models
  • Designed for use with face shields