Learn More
ORS Nasco FACESHLD UVEX BIONIC CL HC /
Catalog No. 502551567
Encompass_Preferred
Honeywell part# S8510. Honeywell Uvex Bionic Face Shields Lens Coating: Anti-fog; Hardcoat Lens Tint: Clear Lens Type: Anti-fog Certifications/Compliance: ANSI Z87.1-2010 CSA Z94.3 Frame Color: Black Lens Material: Polycarbonate Recommended Applications: Safety
Catalog No. 50-255-1567 Supplier ORS Nasco Supplier No. 763S8510
Honeywell part# S8510. Honeywell Uvex Bionic Face Shields Lens Coating: Anti-fog; Hardcoat Lens Tint: Clear Lens Type: Anti-fog Certifications/Compliance: ANSI Z87.1-2010 CSA Z94.3 Frame Color: Black Lens Material: Polycarbonate Recommended Applications: Safety