Learn More
VAI PF-6 FaceVector 3-Ply Masks
Catalog No. 19546531 Shop All VAI Products
Change view
Click to view available options
PF-6 FaceVector 3-Ply Masks; Fastener: 2 Elastic Earloops; Color: Blue; Sterile: No
Catalog No.
19-546-531 PF-6 FaceVector 3-Ply Masks; Fastener: 2 Elastic Earloops; Color: Blue; Sterile: No
1 options
This item is not returnable. View return policy
Catalog No. 19-546-531 Supplier VAI Supplier No. PF6SM2031
Add to Cart
Add to Cart
This item is not returnable. View return policy
PF-6 FaceVector 3-Ply Masks are comprised of 3 layers of polypropylene material with two elastic, silicon-free and lint-free earloop bands.
  • Soft material to help prevent skin irritation or allergies
  • Coated metal strip that acts as a nosepiece to keep the mask securely in place
  • Meets ASTM F2100 Level 1 and EN14683 Type 1 barrier level safety testing parameters.
  • Intended uses include pharmaceutical, biotechnology, lab animal research, compounding pharmacies and hospitals for particle filtration
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Size Standard
Sterility Non-sterile
Disposable Single-use
Model PF-6-SM-2031