Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Facility Traffic Sign - STOP CHECK WITH GUARD BEFORE PROCEEDING

Catalog No. 19169588 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Reflective Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-169-588 Reflective Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-588 Supplier Accuform™ Supplier No. FRR253RA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accuform™ Facility Traffic Signs safely direct and regulate the flow of vehicles and pedestrians around parking areas, dock areas, and roadways.

  • Durable, 0.080 in. aluminum signs will not rust
  • Engineer-Grade Prismatic Reflective sign face provides visibility in low-light conditions
  • UV-resistant inks prevent fading from long-term outdoor use
  • Finishing includes clear UV protective overcoat and post mount holes

Specifications

Grade Engineer
Material Reflective Aluminum
Mounting Type Corner Holes
Languages English
Dimensions (L x W) 24 x 18 in.
Displayed Text STOP CHECK WITH GUARD BEFORE PROCEEDING
Color Red on White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.