Learn More
Amsbio LLC FACSmax Cell Dissociation Solution
Catalog No. 502041288
Encompass
FACSmax™ Cell Dissociation Solution is a chemical separation solution utilizing a cocktail of enzymes carefully calibrated to break up clumped cells. At the same time, FACSmax™ Solution leaves the plasma membrane and intracellular components intact, allowing for maximum cell viability.
Catalog No. 50-204-1288 Supplier Amsbio LLC Supplier No. AMS.T200100
FACSmax™ Cell Dissociation Solution is a chemical separation solution utilizing a cocktail of enzymes carefully calibrated to break up clumped cells. At the same time, FACSmax™ Solution leaves the plasma membrane and intracellular components intact, allowing for maximum cell viability.