Learn More
DIAGNOSTICA STAGO INC Staclot VIIa-rTF
Catalog No. NC1824691
Encompass
Kit for plasma analysis of circulating FVIIa using a clotting method. This unique product is a tool for research studies where the determination of FVIIa levels may be useful due to the fact that it reflects coagulation activation as a result of in vivo FVIIa production.
This item is not returnable. View return policy
Catalog No. NC1824691 Supplier DIAGNOSTICA STAGO INC Supplier No. 00281
Kit for plasma analysis of circulating FVIIa using a clotting method. This unique product is a tool for research studies where the determination of FVIIa levels may be useful due to the fact that it reflects coagulation activation as a result of in vivo FVIIa production.