Learn More
Complement Technology Inc Factor H, 250 μg/vial, frozen liquid, >90% activity vs normal human serum, >95% purity, 1.0 mg/mL concentration, -70°C storage, 155,000 Da molecular weight
Supplier:  Complement Technology Inc A137
Encompass
Factor H, 250 μg/vial, frozen liquid, >90% activity vs normal human serum, >95% purity, 1.0 mg/mL concentration, -70°C storage, 155,000 Da molecular weight
Catalog No. NC0816221