Learn More
Selleck Chemical LLC Factor III-A3078-1MG
SDP
Supplier:  Selleck Chemical LLC A30781MG
Encompass_Preferred
Factor III is humanized form of an antibody capable of targeting tissue factor (coagulation factor F3) It is useful in treating conditions such as tumor progression MW 144 34 KD
Catalog No. 50-313-9170