Learn More
Medchemexpress LLC α-Factor Mating Pheromone, yeast (TFA) | 99.7% | 1798.02 | 10 MG
SDP
Catalog No. 5000422169
Encompass_Preferred
α-Factor Mating Pheromone, yeast TFA is a tridecapeptide secreted by S. cerevisiae α cells via Ste2p receptor. This product is intended for laboratory research and manufacture of substances.
  • Tridecapeptide secreted by S. cerevisiae α cells
  • White to off-white solid appearance
  • Soluble in water at ≥ 100 mg/mL
  • For research use only
Catalog No. 50-004-22169 Supplier Medchemexpress LLC Supplier No. HYP1482A10MG
α-Factor Mating Pheromone, yeast TFA is a tridecapeptide secreted by S. cerevisiae α cells via Ste2p receptor. This product is intended for laboratory research and manufacture of substances.
  • Tridecapeptide secreted by S. cerevisiae α cells
  • White to off-white solid appearance
  • Soluble in water at ≥ 100 mg/mL
  • For research use only