Learn More
Biomatik Corporation Factor Related Apoptosis Ligand (Fasl) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28921
Encompass_Preferred
Factor Related Apoptosis Ligand (Fasl) Polyclonal Antibody is a Rhesus monkey (Simian) Polyclonal Antibody to Factor Related Apoptosis Ligand (FASL). The Factor Related Apoptosis Ligand (Fasl) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rhesus monkey (Simian). Immunogen: Gln102~Leu280. Applications: WB, IHC, ICC, IP. Target Synonyms: Factor Related Apoptosis Ligand (FASL). Accession Number: P63307. Gene ID: 574159. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7585