Learn More
Haematologic Technologies Inc FACTOR VIIA SUBSTRATE
Supplier:  Haematologic Technologies Inc SN17A
Encompass_Preferred
Fluorogenic substrate, Contains fluorescent reporter group 6-amino-1-naphthalene-sulfonamide (ANSN) for monitoring enzyme activity of FACTOR VIIa or THROMBIN, D-FPR-ANSNH-C6H11 • 2 HCl, 1 milligram, RUO, Supplied in DMSO, store at -20 C.
Catalog No. 50-883-409