Learn More
Biocare Medical, LLC Factor VIII Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CP039A
Encompass_Preferred
Factor VIII is a rabbit polyclonal antibody that is intended for laboratory use in the qualitative identification of von Willebrand factor protein by immunohistochemistry (IHC) in formalin-fixed paraffin-embedded (FFPE) human tissues. The clinical interpretation of any staining or its absence should be complemented by morphological studies using proper controls and should be evaluated within the context of the patient clinical history and other diagnostic tests by a qualified pathologist.
Catalog No. 50-827-50