Learn More
Diagnostic Biosystems FACTOR VIII R ANTIGEN Factor VIII Related Antigen/Von Willebrand Factor
Catalog No. 502407601
Encompass_Preferred
This antibody reacts with human factor VIII related antigen. It stains endothelial cells and megakaryocytes in various human tissues. This antibody does not cross-react with any other cell types.
Catalog No. 50-240-7601 Supplier Diagnostic Biosystems Supplier No. PDR014
This antibody reacts with human factor VIII related antigen. It stains endothelial cells and megakaryocytes in various human tissues. This antibody does not cross-react with any other cell types.