Learn More
New England Biolabs, Inc. Factor Xa Protease – 250 µg
SDP
Catalog No. 50811907
Encompass
Factor Xa cleaves after the arginine residue in its preferred cleavage site Ile-Glu/Asp-Gly-Arg. It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate. The most common secondary site, among those that have been sequenced, is Gly-Arg.
Catalog No. 50-811-907 Supplier New England Biolabs, Inc. Supplier No. P8010L
Factor Xa cleaves after the arginine residue in its preferred cleavage site Ile-Glu/Asp-Gly-Arg. It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate. The most common secondary site, among those that have been sequenced, is Gly-Arg.