Learn More
New England Biolabs, Inc. Factor Xa Protease – 50 µg
SDP
Supplier:  New England Biolabs, Inc. P8010S
Encompass
Factor Xa cleaves after the arginine residue in its preferred cleavage site Ile-Glu/Asp-Gly-Arg. It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate. The most common secondary site, among those that have been sequenced, is Gly-Arg.
Catalog No. 50-811-908